Order Entry
Poland
ContactUsLinkComponent
Human Perilipin 3/TIP47 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative de termination of human Perilipin 3/TIP47 in tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA6685-96
Supplier:  ANTIBODIES.COM
New Product
Human Perilipin 3/TIP47 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative de termination of human Perilipin 3/TIP47 in tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA6685-96
Supplier:  ANTIBODIES.COM