Order Entry
Poland
ContactUsLinkComponent
Mouse EKLF/KLF1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinat ion of mouse EKLF/KLF1 in serum, plasma or other biological fluids. 1 * 96 Tests
Catalog # ANTIA6325-96
Supplier:  ANTIBODIES.COM
New Product
Mouse EKLF/KLF1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinat ion of mouse EKLF/KLF1 in serum, plasma or other biological fluids. 1 * 96 Tests
Catalog # ANTIA6325-96
Supplier:  ANTIBODIES.COM