Order Entry
Poland
ContactUsLinkComponent
Mouse gamma Endorphin ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative d etermination of mouse gamma Endorphin in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA7328-48
Supplier:  ANTIBODIES.COM
New Product
Mouse gamma Endorphin ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative d etermination of mouse gamma Endorphin in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA7328-48
Supplier:  ANTIBODIES.COM