Order Entry
Poland
ContactUsLinkComponent
Mouse CACNA1H ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of mouse CACNA1H in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA7200-48
Supplier:  Antibodies.com
New Product
Mouse CACNA1H ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of mouse CACNA1H in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA7200-48
Supplier:  Antibodies.com