Order Entry
Poland
ContactUsLinkComponent
Human Fetal Hemoglobin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative det ermination of human Fetal Hemoglobin in serum, plasma or erythrocyte lysates. 1 * 48 Tests
  ANTIA6655-48
New Product
Human Fetal Hemoglobin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative det ermination of human Fetal Hemoglobin in serum, plasma or erythrocyte lysates. 1 * 48 Tests
  ANTIA6655-48