Order Entry
Poland
ContactUsLinkComponent
Human TOM70 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human TOM70 in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6273-48
Supplier:  Antibodies.com
New Product
Human TOM70 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human TOM70 in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6273-48
Supplier:  Antibodies.com