Order Entry
Poland
ContactUsLinkComponent
Rat HuR/ELAVL1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of rat HuR/ELAVL1 in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA7799-48
Supplier:  Antibodies.com
New Product
Rat HuR/ELAVL1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of rat HuR/ELAVL1 in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA7799-48
Supplier:  Antibodies.com