Order Entry
Poland
ContactUsLinkComponent
Mouse FSH beta ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of mouse FSH beta in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA8090-48
Supplier:  Antibodies.com
New Product
Mouse FSH beta ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of mouse FSH beta in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA8090-48
Supplier:  Antibodies.com