Order Entry
Poland
ContactUsLinkComponent
Human PCTAIRE1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of human PCTAIRE1 in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA7933-48
Supplier:  Antibodies.com
New Product
Human PCTAIRE1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of human PCTAIRE1 in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA7933-48
Supplier:  Antibodies.com