Order Entry
Poland
Orders LinkContactUsLinkComponent
3-(4-Chlorophenoxy)butan-2-one, tech

3-(4-Chlorophenoxy)butan-2-one, tech

Bottle
Catalog # APOSOR33209-5G
New Product
Supplier:   Apollo Scientific
3-(4-Chlorophenoxy)butan-2-one, tech
Catalog # APOSOR33209-5G
Supplier:  Apollo Scientific
New Product
CAS Number:  3782-11-4
3-(4-Chlorophenoxy)butan-2-one, tech

Bottle

CAS Number:  3782-11-4
Price
30 490 zł
Log in for availability
1 * 5 g
Product Details & Documents
  • CAS Number:3782-11-4
  • Formula:C₁₀H₁₁ClO₂
  • MDL Number:MFCD00243668
  • Molecular Weight:198.646 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.