Złóż zamówienie
Poland
Orders LinkContactUsLinkComponent
Bromophenol Red Indicator

Bromophenol Red Indicator

Bottle
Catalog # APOSOR1012563-500G
Supplier:   Apollo Scientific
Bromophenol Red Indicator
Catalog # APOSOR1012563-500G
Supplier:  Apollo Scientific
CAS Number:  
Bromophenol Red Indicator

Bottle

CAS Number:  
Price
5 090 zł
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  4,4'-(3H-2,1-Benzoxathiol-3-ylidene)bis[2-bromophenol] S,S-dioxide
  • Molecular Weight:512,17 g/mol
Specifications

Specifications

  • Rodzaj opakowania:
    Bottle

Specifications