PDMS SPME FIBER (POLYDIMETHYLSILOXANE) #FIBER THICKNESS 30µM, FIBER LENGTH 10MM, #COLOR CODE: GOLDEN, PACK OF 3PCS for TRI Plus SMART 1 * 1 items
Catalog # HYPE36SP03P3-SM
Dostawca: Thermo Fisher Scientific