Drying unit DU 1150 electric For machines 1150 mm wide & optimum internal & external drying of wash items 1 * 1 items
MIEL12678650
: MIELE
New Product
Drying unit DU 1150 electric For machines 1150 mm wide & optimum internal & external drying of wash items 1 * 1 items
MIEL12678650
: MIELE
New Product
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR® Facial Tissues
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection