Multiscreen® High Volume Filter Plates
MultiScreen® High volume filter plates, PP, Opaque, Hydrophilic Durapore®
membrane, 0,45 µm, 96, +
membrane, 0,45 µm, 96, +
Catalog # MILYMVHVN4525
Supplier: MILLIPORE
Jump to:
Product Details & Documents
The original multiwell design is valuable for many applications which require immersing the filter in an assay tray. The easily removable underdrain and unique plate design are ideal for assays such as Elispot, PAMPA, cell screening (migration, invasion or chemotaxis) or for topcount detection users.
- Sample prep
- Protein kinase/phosphatase assays
- Receptor/ligand binding assays
Specifications
- Membrane:Hydrophilic Durapore®
membrane - No. of wells:96
- Pore size:0,45 µm
- Sterile:+
- Colour:Opaque
- Description:MultiScreen® High volume filter plates
- Material:PP
- Seq:1



