Order Entry
Poland
ContactUsLinkComponent
Comfort MTP rack for upright freezers for SBS format (HxD) 4x4=16 plates, 90mm high , stainless steel, dimensions (HxDxW) 369x360x13 5mm 1 * 1 items
Catalog # TENATE35664
Supplier:  TENAK
New Product
Comfort MTP rack for upright freezers for SBS format (HxD) 4x4=16 plates, 90mm high , stainless steel, dimensions (HxDxW) 369x360x13 5mm 1 * 1 items
Catalog # TENATE35664
Supplier:  TENAK