Order Entry
Poland
ContactUsLinkComponent
Comfort MTP racks for chest freezers for 7 SBS format plates, 85mm high, stainless steel, dimensions (HxDxW) 645x135x92mm 1 * 1 items
Catalog # TENATE35288
Supplier:  TENAK
New Product
Comfort MTP racks for chest freezers for 7 SBS format plates, 85mm high, stainless steel, dimensions (HxDxW) 645x135x92mm 1 * 1 items
Catalog # TENATE35288
Supplier:  TENAK