Order Entry
Poland
ContactUsLinkComponent
Standard MTP rack for upright freezers for SBS format (HxD) 2x4=8 plates, 90mm high, stainless steel, dimensions (HxDxW) 185x360x135 mm 1 * 1 items
Catalog # TENATE27660
Supplier:  TENAK
New Product
Standard MTP rack for upright freezers for SBS format (HxD) 2x4=8 plates, 90mm high, stainless steel, dimensions (HxDxW) 185x360x135 mm 1 * 1 items
Catalog # TENATE27660
Supplier:  TENAK