Order Entry
Poland
ContactUsLinkComponent
Standard MTP rack for upright freezers for SBS format (HxD) 5x4=20 plates, 55mm high, stainless steel, dimensions (HxDxW) 286x360x13 5mm 1 * 1 items
Catalog # TENATE27624
Supplier:  TENAK
New Product
Standard MTP rack for upright freezers for SBS format (HxD) 5x4=20 plates, 55mm high, stainless steel, dimensions (HxDxW) 286x360x13 5mm 1 * 1 items
Catalog # TENATE27624
Supplier:  TENAK