Order Entry
Poland
ContactUsLinkComponent
Standard MTP racks for chest freezers for 12 SBS format plates, 55mm high, stainless steel, dimensions (HxDxW) 682x135x92mm 1 * 1 items
Catalog # TENATE27274
Supplier:  TENAK
New Product
Standard MTP racks for chest freezers for 12 SBS format plates, 55mm high, stainless steel, dimensions (HxDxW) 682x135x92mm 1 * 1 items
Catalog # TENATE27274
Supplier:  TENAK