Order Entry
Poland
ContactUsLinkComponent
Comfort rack for chest freezers for 5 cryoboxes, 130mm high, stainless steel, with spring lock, dimensions (HxDxW) 669x140x140mm, de livered including standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
Catalog # TENATE24576B10
Supplier:  TENAK
New Product
Comfort rack for chest freezers for 5 cryoboxes, 130mm high, stainless steel, with spring lock, dimensions (HxDxW) 669x140x140mm, de livered including standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
Catalog # TENATE24576B10
Supplier:  TENAK