Order Entry
Poland
ContactUsLinkComponent
Comfort rack for chest freezers for 12 cryoboxes, 50mm high, stainless steel, with spring lock, dimensions (HxDxW) 664x140x140mm, de livered including standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
  TENATE24516B10
 :  TENAK
New Product
Comfort rack for chest freezers for 12 cryoboxes, 50mm high, stainless steel, with spring lock, dimensions (HxDxW) 664x140x140mm, de livered including standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
  TENATE24516B10
 :  TENAK