Order Entry
Poland
ContactUsLinkComponent
Comfort rack for chest freezers for 10 cryoboxes, 32mm high, stainless steel, with spring lock, dimensions (HxDxW) 359x140x140mm, de livered including standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
Catalog # TENATE24460B10
Supplier:  TENAK
New Product
Comfort rack for chest freezers for 10 cryoboxes, 32mm high, stainless steel, with spring lock, dimensions (HxDxW) 359x140x140mm, de livered including standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
Catalog # TENATE24460B10
Supplier:  TENAK