Order Entry
Poland
ContactUsLinkComponent
Comfort rack for chest freezers for 8 cryoboxes, 32mm high, stainless steel, with spring lock, dimensions (HxDxW) 288x140x140mm, del ivered including standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
  TENATE24458B10
 :  TENAK
New Product
Comfort rack for chest freezers for 8 cryoboxes, 32mm high, stainless steel, with spring lock, dimensions (HxDxW) 288x140x140mm, del ivered including standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
  TENATE24458B10
 :  TENAK