Comfort rack for chest freezers for 12 cryoboxes, 40mm high, stainless steel, with spring lock, dimensions (HxDxW) 508x140x140mm, de livered including standard white cardboard boxes and 9x9 cell dividers 1 * 1 items
TENATE24438B
: TENAK