Order Entry
Poland
ContactUsLinkComponent
Comfort rack for chest freezers for 6 cryoboxes, 40mm high, stainless steel, with spring lock, dimensions (HxDxW) 256x140x140mm, del ivered including standard white cardboard boxes and 9x9 cell dividers 1 * 1 items
Catalog # TENATE24430B
Supplier:  TENAK
New Product
Comfort rack for chest freezers for 6 cryoboxes, 40mm high, stainless steel, with spring lock, dimensions (HxDxW) 256x140x140mm, del ivered including standard white cardboard boxes and 9x9 cell dividers 1 * 1 items
Catalog # TENATE24430B
Supplier:  TENAK