Order Entry
Poland
ContactUsLinkComponent
Comfort rack for upright freezers for (HxD) 7x5=35 cryoboxes, 32mm high, stainless steel, sliding shelves, dimensions (HxDxW) 253x68 3x140mm, delivered including standard white cardboard boxes and 9x9 cell dividers 1 * 1 items
Catalog # TENATE24154B
Supplier:  TENAK
New Product
Comfort rack for upright freezers for (HxD) 7x5=35 cryoboxes, 32mm high, stainless steel, sliding shelves, dimensions (HxDxW) 253x68 3x140mm, delivered including standard white cardboard boxes and 9x9 cell dividers 1 * 1 items
Catalog # TENATE24154B
Supplier:  TENAK