Order Entry
Poland
ContactUsLinkComponent
Comfort rack for upright freezers for (HxD) 4x5=20 cryoboxes, 40mm high, stainless steel, sliding shelves, dimensions (HxDxW) 171x68 3x140mm, delivered including standard white cardboard boxes and 9x9 cell dividers 1 * 1 items
Catalog # TENATE24034B
Supplier:  TENAK
New Product
Comfort rack for upright freezers for (HxD) 4x5=20 cryoboxes, 40mm high, stainless steel, sliding shelves, dimensions (HxDxW) 171x68 3x140mm, delivered including standard white cardboard boxes and 9x9 cell dividers 1 * 1 items
Catalog # TENATE24034B
Supplier:  TENAK