Order Entry
Poland
ContactUsLinkComponent
Classic rack for chest freezers for 17 cryoboxes, 40mm high, stainless steel, with locking rod, dimensions (HxDxW) 718x140x140mm, de livered including standard white cardboard boxes and 9x9 cell dividers 1 * 1 items
  TENATE21389B
 :  TENAK
New Product
Classic rack for chest freezers for 17 cryoboxes, 40mm high, stainless steel, with locking rod, dimensions (HxDxW) 718x140x140mm, de livered including standard white cardboard boxes and 9x9 cell dividers 1 * 1 items
  TENATE21389B
 :  TENAK