Eco AluRack for upright freezers for (HxD) 6x4=24 cryoboxes, 50mm high, aluminium, dimensions (HxDxW) 334x564x139mm, delivered inclu ding standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
TENATE11236B10
: TENAK