NEEDLE SPINAL YALE 25GX3 0,5X75MM 1 * 25 items
Catalog # BDAM405246
Supplier: BD Medical
New Product
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Pipette tips, Finntip™
Supplier: Thermo Fisher Scientific
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Ethanol absolute ≥99.5% USP, Ph. Eur.
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol 96% vol Ph. Eur.
Supplier: VWR Chemicals
VWR® Reagent Reservoirs
Supplier: VWR Collection
VWR® Autoclavable Biohazard Bags, Polypropylene, Orange
Supplier: VWR Collection
Assistent Brushes with Cotton Tip Bristles
Supplier: GLASWARENFABRIK KARL HECHT
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
NEEDLE SPINAL YALE 22GX2,5 0,7X65MM 1 * 25 items
Supplier: BD Medical
NEEDLE SPINAL QUINCKE 25GX2 0,5X50MM 1 * 25 items
Supplier: BD Medical
Combitips® advanced PCR Clean - Positive Displacement Pipette Tips
Supplier: EPPENDORF
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals