Order Entry
Poland
ContactUsLinkComponent
S3s hro ci hi lg fo sr 7 zip boot, water resistant nabutek leather smellstop lining, fortrex puncture resistant patent + dry'n ai r footbed slimcap toe-cap fortrex sole pu + rubber 1 * 1 PAIR
Catalog # PRTWB1611BKR36
Supplier:  Portwest
New Product
S3s hro ci hi lg fo sr 7 zip boot, water resistant nabutek leather smellstop lining, fortrex puncture resistant patent + dry'n ai r footbed slimcap toe-cap fortrex sole pu + rubber 1 * 1 PAIR
Catalog # PRTWB1611BKR36
Supplier:  Portwest