Order Entry
Poland
Orders LinkContactUsLinkComponent
 

hMSC Proliferation Medium, Basal

hMSC proliferation medium, FCS-kit, Liquid, 500 mL
Catalog # PRTV2110901
Supplier:   PROVITRO
hMSC Proliferation Medium, Basal
Catalog # PRTV2110901
Supplier:  PROVITRO
CAS Number:  
hMSC Proliferation Medium, Basal
hMSC Proliferation Medium, Basal

hMSC proliferation medium, FCS-kit, Liquid, 500 mL

CAS Number:  
Price
1 220 zł
Log in for availability
1 * 500 mL
Product Details & Documents

The hMSC Proliferation Medium is a sterile liquid culture medium designed for cultivating human mesenchymal stem cells (hMSC). Delivered as a basal medium, it is well-suited for culturing hMSC upon the addition of supplement mix components.

  • Formulation is optimised for the initial seeding of 6000 cells/cm², supporting growth up to confluence (approximately 90%)
  • Medium does not require feeder layers, matrix substrates, or additional substances for effective cell cultivation

Due to the possibility of reduced proliferative activity, we recommend to use the antibiotic supplement for freshly isolated cells only. Endothelial cell proliferation medium is thoroughly tested after each production. All components are tested in a stringent biological assay.

Each batch is checked for human endothelial cells proliferating characteristics. The cells cultured in endothelial cell growth medium are checked regarding their morphology, the adherence rate, the colony forming efficiency, and the population doubling time.

Storge: Store the supplemented Skeletal muscle cell growth medium in a dark environment at temperatures ranging from 4 to 8 °C for a maximum duration of one month. Avoid heating the medium beyond 37 °C and refrain from using uncontrolled heat sources, such as microwave appliances. When utilising a portion of the medium, extract the required amount from the bottle and warm it appropriately. Never freeze the medium, as this could result in elevated salt concentrations due to the separation of pure water during freezing, leading to irreversible damage.

Specifications

Specifications

  • Media Format:
    Liquid
  • Pk:
    500 mL
  • Description:
    hMSC proliferation medium, FCS-kit
  • Cat. No.:
    PRTV2110901
  • Media Base:
    Basal Medium

Specifications