NEXOPART Test Sieve with stainless steel frame 200X50 mm Nominal size: 32 µ Standard: DIN ISO 3310-1 1 * 1 items
Catalog # HAVE206141163
Supplier: NEXOPART
New Product
NEXOPART Test Sieve with stainless steel frame 200X50 mm Nominal size: 32 µ Standard: DIN ISO 3310-1 1 * 1 items
Catalog # HAVE206141163
Supplier: NEXOPART
New Product
Frequently Bought Together
3-Packs, Research® plus (General Lab Product)
Supplier: EPPENDORF
VWR®, Temperature Indicating Strips
Supplier: VWR Collection
VWR®, Digital Timer, One Channel
Supplier: VWR Collection
Pipette stand, linear
Supplier: Sartorius
Pipette holder, SINGLE™
Supplier: GILSON
VWR®, Digital Refrigerator and Freezer Thermometer
Supplier: VWR Collection
Repetitive pipette, Multipette® E3 / E3x
Supplier: EPPENDORF
VWR®, Environmentally Friendly Thermometers, Easy Read
Supplier: VWR Collection
VWR®, Digital Timer, One Channel
Supplier: VWR Collection
Accessories for PIPETBOY acu / pro
Supplier: Integra
VWR®, Digital Timer, Four Channels, with Clock
Supplier: VWR Collection
Pipette holder 2 and charger shell 2 for stands and carousels
Supplier: EPPENDORF
PRINTER RIBBON CARTRIDGE f. CBM-910II (Citizen) 1 * 1 items
Supplier: SHP Steriltechnik