CONOSTAN CUSTOM BLEND AM SPECIAL 500 PPM 1 * 100 g
Catalog # PROOCON150-250-010
Supplier: LGC Standards PROMOCHEM
New Product
CONOSTAN CUSTOM BLEND AM SPECIAL 500 PPM 1 * 100 g
Catalog # PROOCON150-250-010
Supplier: LGC Standards PROMOCHEM
New Product
Frequently Bought Together
VWR®, Total Nitrogen Standards (CRMs)
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR®, Refractive index standards
Supplier: VWR Chemicals
Buffer solutions in sachets
Supplier: Mettler - Toledo
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
Buffer solutions in sachets, perpHect® Pouches, Orion™
Supplier: Thermo Orion
TOC standard, according to ISO/CEN EN1484, Supelco®
Supplier: Merck
pH Standard Solutions, IUPAC, Certified
Supplier: Hach
pH buffer, single-use, SINGLET™
Supplier: Hach
Reagents for alkalinity determination using Lovibond® MD photometers
Supplier: Lovibond