KWIK -DIFF STAIN KIT 500ML 1 * 1 items
Catalog # SHAN9990700
Supplier: SHANDON
New Product
Frequently Bought Together
Formaldehyde 4% stabilised, buffered (pH 7.0 ±0.1)
Supplier: VWR Chemicals
Test kits, photometric, Nitrite, LCK cuvette tests
Supplier: Hach
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Test kits, nitrite, Spectroquant®, Supelco®
Supplier: Merck
Test strips for copper, MQuant®, Supelco®
Supplier: Merck
Lugol's iodine solution, Sigma-Aldrich®
Supplier: Merck
pH indicator papers, MQuant®, Supelco®
Supplier: Merck
VWR®, OCT Embedding matrix for frozen sections
Supplier: VWR Chemicals
Formalin 10%, Q Path®, buffered
Supplier: VWR Chemicals
Sodium hydroxide, pellets, EMSURE® for analysis, Supelco®
Supplier: Merck
Ammonia solution 25%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Test Strips, nitrate, MQuant®, Supelco®
Supplier: Merck
Test kits, photometric, COD, LCK cuvette tests
Supplier: Hach
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
VWR®, Powder packs for determination of chlorine, reagents for photometers
Supplier: VWR Chemicals