Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Stain, Modified Wright Pack 1 * 5 items
Catalog # SHAN3000
Supplier:
SHANDON
New Product
Expand image
Stain, Modified Wright Pack 1 * 5 items
Catalog # SHAN3000
Supplier:
SHANDON
New Product
Frequently Bought Together
HACH 100 ML SULFIDE 1 (100 TESTS) 1 * 100 mL
Supplier: WATER ID PRIMELAB
30 ML PL DISSOX 2 (75 TESTS) 1 * 30 mL
Supplier: WATER ID PRIMELAB
Cuvette tests for water testing for WTW spectrophotometers and photometers
Supplier: WTW
Powder pillows, reagents for Hach instruments
Supplier: Hach
BIOTECTOR 1.8N SCHWEFELSAURE 80MG/L MN
Supplier: Hach
Reagents for hydrazine determination using Lovibond® MD photometers
Supplier: Lovibond
Reagents for iron determination using Lovibond® MD Photometers
Supplier: Lovibond
[EN]LTBR3-TAG FREE HUMAN 1 * 1 mg
Supplier: RECEPTON
[EN]CD40-FC HUMAN 1 * 10 mg
Supplier: RECEPTON