FORMAL-FIXX PRE-FIILED 125x30ml 1 * 125 items
numer katalogowy SHAN9990914
Dostawca: SHANDON
New Product
FORMAL-FIXX PRE-FIILED 125x30ml 1 * 125 items
numer katalogowy SHAN9990914
Dostawca: SHANDON
New Product
Frequently Bought Together
Eppendorf® Protein LoBind® Microcentrifuge Tubes
Dostawca: EPPENDORF
Acetone ≥99%, TECHNICAL
Dostawca: VWR Chemicals
VWR® Standard Line Transfer Pipettes
Dostawca: VWR Collection
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Dostawca: VWR Chemicals
Safe-Lock Tubes, 1,5 ml - Microtubes
Dostawca: EPPENDORF
VWR®, Transfer Pipettes, High Performance
Dostawca: VWR Collection
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Dostawca: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Dostawca: VWR Chemicals
Water, AnalaR NORMAPUR® ISO 3696 Grade 3 analytical reagent
Dostawca: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Dostawca: VWR Chemicals
Safe-Lock Tubes, 2,0 ml - Microtubes
Dostawca: EPPENDORF
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Dostawca: Merck
VWR® Serological Pipette, Standard Line
Dostawca: VWR Collection
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Dostawca: VWR Chemicals