0 zł
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Poland
ContactUsLinkComponent
GLYO-FIXX CONCENTRATE, 3800ML 1 * 1 items
SHAN6764265
:
SHANDON
New Product
GLYO-FIXX CONCENTRATE, 3800ML 1 * 1 items
SHAN6764265
:
SHANDON
New Product
Frequently Bought Together
Schiff reagent for histology, Sigma-Aldrich®
: Merck
Test kits, COD cell test, Spectroquant®, Supelco®
: Merck
Test kits, nitrite, Spectroquant®, Supelco®
: Merck
Test kits, surfactants, Spectroquant®, Supelco®
: Merck
Test kits, nitrogen (total) cell tests, Spectroquant®, Supelco®
: Merck
Test kits, ammonium, Spectroquant®, Supelco®
: Merck
pH indicator strips, MQuant®, Supelco®
: Merck
Test kits, photometric, Zinc, LCK and LCS cuvette tests
: Hach
Papanicolaou's solution 2a Orange G (OG 6), Sigma-Aldrich®
: Merck
Test kits, phosphate, Spectroquant®, Supelco®
: Merck
Test Strips, nitrate, MQuant®, Supelco®
: Merck
Sodium hydroxide 35% in aqueous solution, TECHNICAL
: VWR Chemicals
Test kits, nitrate, Spectroquant®, Supelco®
: Merck
Test kits, photometric, COD, LCK cuvette tests
: Hach
Formalin 10%, Q Path®, buffered
: VWR Chemicals