DECAL 6x0.94L BOTTLE 1 * 6 items
Catalog # SHAN8340
Supplier: SHANDON
New Product
Frequently Bought Together
Ethanol eurodenatured ≥96%, GPR RECTAPUR® (1:1:1)
Supplier: VWR Chemicals
Paraffin, liquid, liquid Ph. Eur.
Supplier: VWR Chemicals
Acetone ≥99.7%, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, CHROMASOLV™, gradient grade for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Toluene ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Toluene, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
n-Hexane ≥97%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Isoamyl alcohol, pure analytical reagent
Supplier: CHEMPUR
ETHANOL 70% SOLUTION 1 * 1 L
Supplier: CHEMPUR
Acetonitrile ≥99.9% for HPLC, for gradient grade far UV
Supplier: Thermo Fisher Scientific
Caffeic acid, Supelco®
Supplier: Merck
Acetonitrile, extra dry over Molecular Sieve 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific
Ethanol absolute, anhydrous (max. 0.003% H₂O) ≥99.8%
Supplier: VWR Chemicals