HISTOPLAST LP 8 x 1Kg 1 * 8 items
Catalog # SHAN8332
Supplier: SHANDON
New Product
Frequently Bought Together
Dichloromethane, anhydrous ≥99.7% stabilised
Supplier: Thermo Fisher Scientific
Ethyl acetate ≥99.5% for HPLC
Supplier: Thermo Fisher Scientific
ETHANOL 96% SOLUTION 1 * 250 mL
Supplier: CHEMPUR
HAEMATOXYLIN GILL 3 3800ML 1 * 1 items
Supplier: SHANDON
Bromoform 96% stabilised
Supplier: Thermo Fisher Scientific
11 components: Ca 2mg/l Zr 5mg/l K 5mg/l Zn 5mg/l Na 5mg/l Fe 5mg/l Al 5mg/l Ti 5mg/l Si 5mg/l P 10mg/l Mn 5mg/l in HNO3 5% 1 * 1 L
Supplier: CUSTOM MADE CHEMICALS LAB
Cetrimonium bromide, Supelco®
Supplier: Merck
Dichloromethane 99.8% stabilised for HPLC
Supplier: Thermo Fisher Scientific
Tetrachloroethylene ≥99%
Supplier: Thermo Fisher Scientific
Ethanol absolute Ph. Eur., J.T.Baker®
Supplier: Avantor
Tetrahydrofuran
Supplier: Merck
2-Methylbutane ≥99%, Reagent Grade
Supplier: Honeywell Chemicals
N,N-Dimethylacetamide, extra dry 99.5%, AcroSeal®
Supplier: Thermo Fisher Scientific