ATORVASTATIN EPOXY TETRAHYDROFURAN ANALOG (EP IMPURITY D2) 1 * 50 mg
Catalog # MSPPSZA037055-50MG
Supplier: MARKET SOURCE PART PROCESS
ATORVASTATIN EPOXY TETRAHYDROFURAN ANALOG (EP IMPURITY D2) 1 * 50 mg
Catalog # MSPPSZA037055-50MG
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together
Sodium chloride ≥98%, TECHNICAL
Supplier: VWR Chemicals
Magnesium sulphate, (dried basis) ≥98%, GPR RECTAPUR®
Supplier: VWR Chemicals
Ammonium acetate ≥98.0%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ammonium sulphate ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Potassium iodide ≥99.5%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Sodium hydrogen carbonate 99.7-100.3%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Barium chloride dihydrate ≥99.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Sodium carbonate ≥97%, TECHNICAL
Supplier: VWR Chemicals