ATORVASTATIN RELATED COMP.C CALCIUM SALT 1 * 50 mg
Catalog # MSPPSZA037004-50MG
Supplier: MARKET SOURCE PART PROCESS
ATORVASTATIN RELATED COMP.C CALCIUM SALT 1 * 50 mg
Catalog # MSPPSZA037004-50MG
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together
Sodium hydrogen carbonate ≥98%, TECHNICAL
Supplier: VWR Chemicals
Sodium carbonate, anhydrous 99.5-100.5% NF, Ph. Eur., BP
Supplier: VWR Chemicals
Potassium chloride 99.5-101.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Potassium dihydrogen phosphate ≥99%, powder, GPR RECTAPUR®
Supplier: VWR Chemicals
Sodium chloride ≥98%, TECHNICAL
Supplier: VWR Chemicals
Ammonium hydrogen carbonate, LiChropur™ for LC-MS, Supelco®
Supplier: Merck
Sodium salicylate, EMSURE® for analysis, Supelco®
Supplier: Merck
Sodium peroxodisulphate, EMSURE® for analysis, Supelco®
Supplier: Merck
Sodium hydrogen carbonate ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Potassium dihydrogen phosphate ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Potassium iodide 99.0-100.5% Ph. Eur.
Supplier: VWR Chemicals
Calcium chloride dihydrate 99.0-103.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Disodium hydrogen phosphate dodecahydrate ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals