Order Entry
Poland
Orders LinkContactUsLinkComponent
ReadiPrep™ Exosome Isolation Kit
ReadiPrep™ Exosome Isolation Kit
50 preparations, 50
Catalog # AATB60204
Supplier:   AAT Bioquest
ReadiPrep™ Exosome Isolation Kit
Catalog # AATB60204
Supplier:  AAT Bioquest
CAS Number:  
ReadiPrep™ Exosome Isolation Kit
50 preparations, 50
CAS Number:  
Price
1 150 zł
Log in for availability
1 * 50 SZT
Product Details & Documents
Designed for consistent and dependable results. The ReadiPrep™ exosome isolation kit offers a streamlined and effective approach to isolating intact exosomes from diverse biological samples, including cell culture media and serum.
  • Robust protocol
  • Produced exosomes are suitable for further downstream analyses or purification via affinity-based methods
  • Six-month shelf life
The principle behind this kit is based on the selective precipitation of exosomes by inducing the removal of water molecules from the solution.
Delivery information: Shipped at RT.
Caution: Research use only.
Specifications
Specifications
  • Pk:
    50
  • Pk Info:
    50 preparations