Order Entry
Poland
ContactUsLinkComponent
Biotinylated Human HLA-C 0303,B2M Monomer Protein HLM-H82E9 is expressed from human 293 cells HEK293. It contains AA Gly 25- Ile 308 HLA-C 0303, Ile 21-Met 119 B2M Accession QDK63961.1 HLA-C 0303, P61769-1 B2M. 1 * 200 µG
Catalog # ACRBHLM-H82E9-200
Supplier:  ACRO Biosystems
Biotinylated Human HLA-C 0303,B2M Monomer Protein HLM-H82E9 is expressed from human 293 cells HEK293. It contains AA Gly 25- Ile 308 HLA-C 0303, Ile 21-Met 119 B2M Accession QDK63961.1 HLA-C 0303, P61769-1 B2M. 1 * 200 µG
Catalog # ACRBHLM-H82E9-200
Supplier:  ACRO Biosystems
Frequently Bought Together