Order Entry
Poland
ContactUsLinkComponent
Biotinylated Human HLA-A 0201,B2M Monomer Protein HLM-H82W3 is expressed from human 293 cells HEK293. It contains AA Gly 25 - Ile 30 8 HLA-A 0201, Ile 21 - Met 119 B2M Accession AAA59606.1 HLA-A 0201, P61769 B2M. 1 * 200 µG
Catalog # ACRBHLM-H82W3-200
Dostawca:  ACRO Biosystems
Biotinylated Human HLA-A 0201,B2M Monomer Protein HLM-H82W3 is expressed from human 293 cells HEK293. It contains AA Gly 25 - Ile 30 8 HLA-A 0201, Ile 21 - Met 119 B2M Accession AAA59606.1 HLA-A 0201, P61769 B2M. 1 * 200 µG
Catalog # ACRBHLM-H82W3-200
Dostawca:  ACRO Biosystems
Frequently Bought Together