Order Entry
Poland
ContactUsLinkComponent
Biotinylated Human HLA-A 3303,B2M Monomer Protein HLM-H82W7 is expressed from human 293 cells HEK293. It contains AA Gly 25 - Ile 30 8 HLA-A 3303, Ile 21 - Met 119 B2M Accession AAA79865.1 HLA-A 3303, P61769-1 B2M. 1 * 200 µG
  ACRBHLM-H82W7-200
Biotinylated Human HLA-A 3303,B2M Monomer Protein HLM-H82W7 is expressed from human 293 cells HEK293. It contains AA Gly 25 - Ile 30 8 HLA-A 3303, Ile 21 - Met 119 B2M Accession AAA79865.1 HLA-A 3303, P61769-1 B2M. 1 * 200 µG
  ACRBHLM-H82W7-200
Frequently Bought Together