Order Entry
Poland
ContactUsLinkComponent
Human MICB Protein, His Tag MIB-H52H3 is expressed from human 293 cells HEK293. It contains AA Ala 23 - Thr 308 Accession NP_005922 .2. Acts as a stress-induced self-antigen that is recognized by gamma delta T cells. Ligand for the KLRK1/NKG2D recept 1 * 100 µG
Catalog # ACRBMIB-H52H3-100
Supplier:  ACROBIOSYSTEMS
Human MICB Protein, His Tag MIB-H52H3 is expressed from human 293 cells HEK293. It contains AA Ala 23 - Thr 308 Accession NP_005922 .2. Acts as a stress-induced self-antigen that is recognized by gamma delta T cells. Ligand for the KLRK1/NKG2D recept 1 * 100 µG
Catalog # ACRBMIB-H52H3-100
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together