RNase Ⅲ 2U/μl 1 * 1.000 AU
Catalog # ACRBRI3-E5143-1000
Supplier: ACROBIOSYSTEMS
Frequently Bought Together
(±)-Tetrahydrofurfuryl alcohol, Sigma-Aldrich®
Supplier: Merck
Benzyl alcohol ≥99.0% analytical reagent
Supplier: VWR Chemicals
Acetone, LiChrosolv® for HPLC, Supelco®
Supplier: Merck
2,5-Di-tert-butylhydroquinone for synthesis, Sigma-Aldrich®
Supplier: Merck
Acetone, Technical Grade
Supplier: CHEMPUR
Phenyl isocyanate, Supelco®
Supplier: Merck
N-Methyl-2-pyrrolidone (NMP) 99.5% for analysis
Supplier: Thermo Fisher Scientific
Methanol, anhydrous ≥99.9%
Supplier: Thermo Fisher Scientific
Acetone (Reag. Ph. Eur.) for analysis, ACS, ISO 1 * 2,5 L
Supplier: PanReac AppliChem
1,2-Dichloroethane, extra dry over Molecular Sieve 99.5%, AcroSeal®
Supplier: Thermo Fisher Scientific
1-Propanol, anhydrous ≥99.9%
Supplier: Thermo Fisher Scientific
Methyl ethyl ketone, extra dry 99.5%, AcroSeal®
Supplier: Thermo Fisher Scientific