Order Entry
Poland
ContactUsLinkComponent
Biotinylated Human HLA-B 0801,B2M Monomer Protein HLM-H82En is expressed from human 293 cells HEK293. It contains AA Gly 25-Val 309 HLA-B 0801, Ile 21-Met 119 B2M Accession BAA22205.1 HLA-B 0801, P61769 B2M. 1 * 25 µG
Catalog # ACRBHLM-H82EN-25UG
Supplier:  ACRO Biosystems
Biotinylated Human HLA-B 0801,B2M Monomer Protein HLM-H82En is expressed from human 293 cells HEK293. It contains AA Gly 25-Val 309 HLA-B 0801, Ile 21-Met 119 B2M Accession BAA22205.1 HLA-B 0801, P61769 B2M. 1 * 25 µG
Catalog # ACRBHLM-H82EN-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together