Order Entry
Poland
ContactUsLinkComponent
Biotinylated Human HLA-B 4001,B2M Monomer Protein HLM-H82Eu is expressed from human 293 cells HEK293. It contains AA Gly 25 - Val 30 9 HLA-B 4001, Ile 21 - Met 119 B2M Accession AAA03719.1 HLA-B 4001, P61769 B2M. 1 * 25 µG
  ACRBHLM-H82EU-25UG
Biotinylated Human HLA-B 4001,B2M Monomer Protein HLM-H82Eu is expressed from human 293 cells HEK293. It contains AA Gly 25 - Val 30 9 HLA-B 4001, Ile 21 - Met 119 B2M Accession AAA03719.1 HLA-B 4001, P61769 B2M. 1 * 25 µG
  ACRBHLM-H82EU-25UG
Frequently Bought Together